Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075274.2 | NP_001068742.1 | 2 - 3 | 336 | 95 | |
GenBank mRNA | AM930551.2 | - | 2 - 3 | 588 | 95 | |
BC118355.1 | - | 2 - 3 | 337 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |