Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075277.1 | NP_001068745.1 | 6 - 7 | 1297 | 74 | |
GenBank mRNA | BC116062.1 | - | 6 - 7 | 1297 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |