Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075291.2 | NP_001068759.1 | 3 - 4 | 469 | 131 | |
GenBank mRNA | BC118120.1 | - | 3 - 4 | 470 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |