Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075479.1 | NP_001068947.1 | 7 - 8 | 781 | 155 | |
GenBank mRNA | BC116035.1 | - | 7 - 8 | 781 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |