Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075548.1 | NP_001069016.1 | 2 - 3 | 326 | 91 | |
GenBank mRNA | BT025410.1 | - | 2 - 3 | 326 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |