Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075566.1 | NP_001069034.1 | 6 - 7 | 674 | 153 | |
GenBank mRNA | BC114781.1 | - | 6 - 7 | 674 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |