Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075632.1 | NP_001069100.1 | 5 - 6 | 1749 | 113 | |
GenBank mRNA | BC116156.1 | - | 3 - 4 | 631 | 113 | |
BC118095.1 | - | 5 - 6 | 1749 | 113 | ||
BC123802.1 | - | 2 - 3 | 1271 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |