Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075652.2 | NP_001069120.2 | 4 - 5 | 748 | 91 | |
GenBank mRNA | BC114778.1 | - | 4 - 5 | 749 | 91 | |
DV822960.1 | - | 4 - 5 | 675 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |