Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075683.2 | NP_001069151.1 | 2 - 3 | 292 | 62 | |
GenBank mRNA | BC118409.1 | - | 2 - 3 | 293 | 62 |
Gene Symbol: LOC533308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243316.1 | NP_001230245.1 | 1 | 240 | 62 | |
GenBank mRNA | BC149555.1 | - | 1 | 241 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |