Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075785.1 | NP_001069253.1 | 3 - 4 | 823 | 85 | |
GenBank mRNA | BC116055.1 | - | 3 - 4 | 823 | 85 | |
GQ166650.1 | - | 1 - 2 | 293 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |