Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075803.2 | NP_001069271.1 | 6 - 7 | 932 | 121 | |
GenBank mRNA | BC118450.1 | - | 6 - 7 | 933 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |