Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075804.2 | NP_001069272.1 | 7 - 8 | 1036 | 64 | |
GenBank mRNA | BC114881.1 | - | 7 - 8 | 1037 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |