Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076013.2 | NP_001069481.1 | 5 - 6 | 409 | 84 | |
GenBank mRNA | BC116151.1 | - | 5 - 6 | 410 | 84 | |
GQ367549.1 | - | 5 - 6 | 398 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |