Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076015.2 | NP_001069483.1 | 6 - 7 | 971 | 139 | |
GenBank mRNA | BC118356.1 | - | 6 - 7 | 972 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |