Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076048.1 | NP_001069516.1 | 3 - 4 | 222 | 75 | |
GenBank mRNA | BC116053.1 | - | 3 - 4 | 222 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |