Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MACROH2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076086.1 | NP_001069554.1 | 7 - 8 | 953 | 71 | |
GenBank mRNA | BC116136.1 | - | 7 - 8 | 953 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |