Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4HTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076115.2 | NP_001069583.1 | 7 - 8 | 1155 | 84 | |
GenBank mRNA | BC114843.1 | - | 7 - 8 | 1156 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |