Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4HTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076115.2 | NP_001069583.1 | 8 - 9 | 1280 | 79 | |
GenBank mRNA | BC114843.1 | - | 8 - 9 | 1281 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |