Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076124.2 | NP_001069592.1 | 1 - 2 | 291 | 63 | |
GenBank mRNA | BC114011.1 | - | 1 - 2 | 292 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |