Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076132.1 | NP_001069600.1 | 3 - 4 | 1180 | 90 | |
GenBank mRNA | BC118219.1 | - | 3 - 4 | 1180 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |