Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076328.1 | NP_001069796.1 | 4 - 5 | 298 | 107 | |
GenBank mRNA | BC118376.1 | - | 4 - 5 | 298 | 107 | |
BT026143.1 | - | 4 - 5 | 277 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |