Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076449.1 | NP_001069917.1 | 3 - 4 | 293 | 146 | |
GenBank mRNA | BC118163.1 | - | 3 - 4 | 293 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |