Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076496.1 | NP_001069964.1 | 3 - 4 | 580 | 100 | |
GenBank mRNA | BC118258.1 | - | 3 - 4 | 580 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |