Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076496.1 | NP_001069964.1 | 5 - 6 | 781 | 113 | |
GenBank mRNA | BC118258.1 | - | 5 - 6 | 781 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |