Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076955.1 | NP_001070423.1 | 6 - 7 | 945 | 122 | |
GenBank mRNA | BC116079.1 | - | 6 - 7 | 945 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |