Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076977.2 | NP_001070445.1 | 1 - 2 | 65 | 79 | |
GenBank mRNA | AM075813.1 | - | 1 - 2 | 66 | 79 | |
BC142038.1 | - | 1 - 2 | 109 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |