Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077019.1 | NP_001070487.1 | 6 - 7 | 606 | 70 | |
GenBank mRNA | BC105559.1 | - | 6 - 7 | 606 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |