Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078129.2 | NP_001071597.1 | 3 - 4 | 244 | 76 | |
GenBank mRNA | BC105388.1 | - | 3 - 4 | 245 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |