Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080349.2 | NP_001073818.1 | 1 - 2 | 105 | 117 | |
GenBank mRNA | BC118400.1 | - | 1 - 2 | 106 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |