Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174062.4 | NP_776487.1 | 1 | 330 | 64 | |
GenBank mRNA | AB003093.1 | - | 1 - 2 | 270 | 64 | |
AB098915.1 | - | 1 - 2 | 132 | 64 | ||
AB098993.1 | - | 1 - 2 | 318 | 64 | ||
AB099065.2 | - | 1 - 2 | 326 | 64 | ||
AF540563.1 | - | 1 - 2 | 78 | 64 | ||
AY911329.1 | - | 1 - 2 | 330 | 64 | ||
BC105376.1 | - | 1 | 331 | 64 | ||
BC151549.1 | - | 1 - 2 | 332 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |