Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174446.3 | NP_776871.2 | 1 - 2 | 99 | 62 | |
GenBank mRNA | BC118278.1 | - | 1 - 2 | 99 | 62 | |
M19885.1 | - | 1 - 2 | 35 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |