Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174446.3 | NP_776871.2 | 4 - 5 | 575 | 59 | |
GenBank mRNA | BC118278.1 | - | 4 - 5 | 575 | 59 | |
M19885.1 | - | 4 - 5 | 511 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |