Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183364.3 | NP_899208.2 | 2 - 3 | 372 | 112 | |
GenBank mRNA | AY279358.1 | - | 2 - 3 | 388 | 112 | |
BC118357.1 | - | 2 - 3 | 373 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |