Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAICS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205813.2 | NP_991382.2 | 1 - 2 | 230 | 124 | |
GenBank mRNA | AY344127.1 | - | 2 - 3 | 106 | 124 | |
BC113344.1 | - | 1 - 2 | 230 | 124 | ||
BT030591.2 | - | 1 - 2 | 72 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |