Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014905.2 | NP_001014905.2 | 6 - 7 | 1045 | 71 | |
GenBank mRNA | BC148053.1 | - | 6 - 7 | 1069 | 71 | |
BT020649.1 | - | 6 - 7 | 1084 | 71 | ||
BT020652.1 | - | 6 - 7 | 1078 | 71 | ||
BT020813.1 | - | 6 - 7 | 1058 | 71 | ||
BT020880.1 | - | 6 - 7 | 1072 | 71 | ||
BT026157.1 | - | 6 - 7 | 1045 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |