Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034739.2 | NP_001029911.1 | 2 - 3 | 158 | 96 | |
GenBank mRNA | BC119865.1 | - | 2 - 3 | 158 | 96 | |
BT021188.1 | - | 2 - 3 | 158 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |