Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037100.3 | NP_001032177.1 | 1 - 2 | 604 | 96 | |
GenBank mRNA | AB195549.1 | - | 1 - 2 | 429 | 96 | |
BC123574.1 | - | 1 - 2 | 605 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |