Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038198.2 | NP_001033287.1 | 3 - 4 | 1063 | 73 | |
GenBank mRNA | AY644769.1 | - | 1 - 2 | 844 | 73 | |
AY644770.1 | - | 1 - 2 | 847 | 73 | ||
BC123390.1 | - | 3 - 4 | 1063 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |