Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040478.2 | NP_001035568.2 | 2 - 3 | 929 | 65 | |
GenBank mRNA | AB110599.1 | - | 2 - 3 | 715 | 65 | |
AB257293.1 | - | 2 - 3 | 126 | 65 | ||
AF093675.1 | - | 2 - 3 | 211 | 65 | ||
BC120454.1 | - | 2 - 3 | 929 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |