Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075191.1 | NP_001068659.1 | 4 - 5 | 1044 | 129 | |
GenBank mRNA | BC120380.1 | - | 4 - 5 | 1044 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |