Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075206.1 | NP_001068674.1 | 5 - 6 | 930 | 84 | |
GenBank mRNA | BT026224.1 | - | 5 - 6 | 930 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |