Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075206.1 | NP_001068674.1 | 6 - 7 | 1127 | 92 | |
GenBank mRNA | BT026224.1 | - | 6 - 7 | 1127 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |