Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075252.2 | NP_001068720.1 | 2 - 3 | 113 | 71 | |
GenBank mRNA | BC120130.1 | - | 2 - 3 | 114 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |