Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPM6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075309.1 | NP_001068777.1 | 5 - 6 | 979 | 123 | |
GenBank mRNA | BC119902.1 | - | 5 - 6 | 979 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |