Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075325.1 | NP_001068793.1 | 4 - 5 | 542 | 82 | |
GenBank mRNA | BC120079.1 | - | 4 - 5 | 542 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |