Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075372.2 | NP_001068840.1 | 2 - 3 | 226 | 91 | |
GenBank mRNA | BC122774.1 | - | 2 - 3 | 227 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |