Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075523.1 | NP_001068991.1 | 6 - 7 | 1183 | 118 | |
GenBank mRNA | BC119958.1 | - | 6 - 7 | 1183 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |