Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075551.1 | NP_001069019.1 | 8 - 9 | 978 | 68 | |
GenBank mRNA | BC126750.1 | - | 8 - 9 | 969 | 68 | |
DQ435444.1 | - | 8 - 9 | 978 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |