Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERPUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075558.1 | NP_001069026.1 | 8 - 9 | 1673 | 102 | |
GenBank mRNA | BC120015.1 | - | 8 - 9 | 1673 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |