Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075561.4 | NP_001069029.2 | 3 - 4 | 438 | 130 | |
GenBank mRNA | BC119854.1 | - | 3 - 4 | 421 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |